Learn More
Selleck Chemical LLC Erythromycin Cyclocarbonate
SDP
Catalog No. 5000002930
Encompass_Preferred
Erythromycin Cyclocarbonate(Davercin) derivative of Erythromycin inhibits protein synthesis of bacteria by binding to the 50S ribosome
Catalog No. 50-000-02930 Supplier Selleck Chemical LLC Supplier No. S75505MG
Erythromycin Cyclocarbonate(Davercin) derivative of Erythromycin inhibits protein synthesis of bacteria by binding to the 50S ribosome