Learn More
Medchemexpress LLC Erythromycin-d3 | 959119-26-7 | 95.0% | 1 MG
SDP
Catalog No. 5000407187
Encompass_Preferred
Erythromycin-d3 is a deuterium-labeled macrolide antibiotic produced by the actinomycete Streptomyces erythreus. It exhibits broad-spectrum antimicrobial activity by binding to bacterial 50S ribosomal subunits, thereby inhibiting RNA-dependent protein synthesis. Beyond its antibiotic properties, Erythromycin also demonstrates antitumor and neuroprotective effects in various research areas.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07187 Supplier Medchemexpress LLC Supplier No. HYB0220S21MG
Erythromycin-d3 is a deuterium-labeled macrolide antibiotic produced by the actinomycete Streptomyces erythreus. It exhibits broad-spectrum antimicrobial activity by binding to bacterial 50S ribosomal subunits, thereby inhibiting RNA-dependent protein synthesis. Beyond its antibiotic properties, Erythromycin also demonstrates antitumor and neuroprotective effects in various research areas.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.