Learn More
Medchemexpress LLC Erythromycin-d6 | 959119-25-6 | 1 MG
SDP
Catalog No. 5000406122
Encompass_Preferred
Erythromycin-d6 is the deuterium labeled version of Erythromycin, a macrolide antibiotic produced by the actinomycete Streptomyces erythreus. Known for its broad spectrum of antimicrobial activity, it functions by binding to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06122 Supplier Medchemexpress LLC Supplier No. HYB0220S1MG
Erythromycin-d6 is the deuterium labeled version of Erythromycin, a macrolide antibiotic produced by the actinomycete Streptomyces erythreus. Known for its broad spectrum of antimicrobial activity, it functions by binding to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS