Learn More
Selleck Chemical LLC Erythromycin estolate
SDP
Catalog No. 5000003241
Encompass_Preferred
Erythromycin (Lubomycine B) a naturally occurring macrolide is derived from Streptomyces erythrus It inhibits bacterial protein synthesis by binding to bacterial 50S ribosomal subunits
Catalog No. 50-000-03241 Supplier Selleck Chemical LLC Supplier No. S553225MG
Erythromycin (Lubomycine B) a naturally occurring macrolide is derived from Streptomyces erythrus It inhibits bacterial protein synthesis by binding to bacterial 50S ribosomal subunits