Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythromycin estolate | 3521-62-8 | 10 MM * 1 ML
SDP

Catalog No. 5000418077
Encompass_Preferred

Erythromycin estolate is a macrolide antibiotic and a derivative of Erythromycin used to treat a wide variety of bacterial infections. It is intended for research use only and not sold to patients. This compound can cause liver injury, primarily cholestatic hepatitis, due to its inhibitory effect on bile acid transport.

  • Purity: 98.0%
  • Molecular weight: 1056.39
  • Formula: C52H97NO18S
  • Functions as a macrolide antibiotic
  • Used to treat bacterial infections
  • Inhibits various flaviviruses including Zika, dengue, and yellow fever
  • Inactivates viral particles by destroying viral membrane integrity
  • Inhibits human BSEP with an IC50 of 4.1 μM
  • Available as a solid, white to off-white in color
  • Storage conditions: 4°C in sealed storage, away from moisture; in solvent: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-18077 Supplier Medchemexpress LLC Supplier No. HYN712110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erythromycin estolate is a macrolide antibiotic and a derivative of Erythromycin used to treat a wide variety of bacterial infections. It is intended for research use only and not sold to patients. This compound can cause liver injury, primarily cholestatic hepatitis, due to its inhibitory effect on bile acid transport.

  • Purity: 98.0%
  • Molecular weight: 1056.39
  • Formula: C52H97NO18S
  • Functions as a macrolide antibiotic
  • Used to treat bacterial infections
  • Inhibits various flaviviruses including Zika, dengue, and yellow fever
  • Inactivates viral particles by destroying viral membrane integrity
  • Inhibits human BSEP with an IC50 of 4.1 μM
  • Available as a solid, white to off-white in color
  • Storage conditions: 4°C in sealed storage, away from moisture; in solvent: -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.