Learn More
Medchemexpress LLC Erythromycin estolate | 3521-62-8 | 100 MG
SDP
Catalog No. 5000419771
Encompass_Preferred
Erythromycin estolate is a macrolide antibiotic derived from Erythromycin, utilized for treating a broad spectrum of bacterial infections. It has been noted to cause liver injury, often presenting as cholestatic hepatitis, with its toxicity linked to inhibiting bile acid transport. This compound also exhibits antiviral properties by inactivating viral particles and destroying their membrane integrity.
  • Used as a macrolide antibiotic.
  • Effective against a wide variety of bacterial infections.
  • Causes liver injury, predominantly cholestatic hepatitis.
  • Toxicity is associated with inhibiting bile acid transport.
  • Inactivates viral particles by destroying viral membrane integrity.
  • Inhibits various flaviviruses, including Zika, dengue, and yellow fever viruses.
Catalog No. 50-004-19771 Supplier Medchemexpress LLC Supplier No. HYN7121100MG
Erythromycin estolate is a macrolide antibiotic derived from Erythromycin, utilized for treating a broad spectrum of bacterial infections. It has been noted to cause liver injury, often presenting as cholestatic hepatitis, with its toxicity linked to inhibiting bile acid transport. This compound also exhibits antiviral properties by inactivating viral particles and destroying their membrane integrity.
  • Used as a macrolide antibiotic.
  • Effective against a wide variety of bacterial infections.
  • Causes liver injury, predominantly cholestatic hepatitis.
  • Toxicity is associated with inhibiting bile acid transport.
  • Inactivates viral particles by destroying viral membrane integrity.
  • Inhibits various flaviviruses, including Zika, dengue, and yellow fever viruses.