Learn More
Apexbio Technology LLC Erythromycin Ethylsuccinate 1264-62-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090017
Encompass_Preferred
Erythromycin Ethylsuccinate (CAS 1264-62-6) is a small-molecule inhibitor targeting the 50S subunit of the bacterial ribosome It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth Erythromycin Ethylsuccinate exerts its biological activity primarily through reversible attachment to the 50S ribosomal subunit resulting in the inhibition of protein biosynthesis In in vitro studies Erythromycin Ethylsuccinate demonstrates inhibitory activity against susceptible bacterial strains with IC50 values typically within the low micromolar range depending on organism type and experimental conditions Based on these pharmacological properties Erythromycin Ethylsuccinate holds research potential in studying ribosome-mediated translation processes antibiotic resistance mechanisms and bacterial physiology
Catalog No. 50-000-90017 Supplier Apexbio Technology LLC Supplier No. B19405.1
Erythromycin Ethylsuccinate (CAS 1264-62-6) is a small-molecule inhibitor targeting the 50S subunit of the bacterial ribosome It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth Erythromycin Ethylsuccinate exerts its biological activity primarily through reversible attachment to the 50S ribosomal subunit resulting in the inhibition of protein biosynthesis In in vitro studies Erythromycin Ethylsuccinate demonstrates inhibitory activity against susceptible bacterial strains with IC50 values typically within the low micromolar range depending on organism type and experimental conditions Based on these pharmacological properties Erythromycin Ethylsuccinate holds research potential in studying ribosome-mediated translation processes antibiotic resistance mechanisms and bacterial physiology