Learn More
Medchemexpress LLC Erythromycin Ethylsuccinate | 1264-62-6 | MFCD00084692 | 79.0% | 1 ML
SDP
Catalog No. 5000409111
Encompass_Preferred
Erythromycin Ethylsuccinate is an antibiotic effective against a variety of bacterial infections. Its antimicrobial spectrum is similar to or slightly broader than that of penicillin. Additionally, it exhibits antiviral activity against HIV-1.
  • Antibiotic useful for treating bacterial infections.
  • Antimicrobial spectrum similar to or slightly wider than penicillin.
  • Has antiviral activity against HIV-1.
  • For research use only.
Catalog No. 50-004-09111 Supplier Medchemexpress LLC Supplier No. HYB095710MM/1ML
Erythromycin Ethylsuccinate is an antibiotic effective against a variety of bacterial infections. Its antimicrobial spectrum is similar to or slightly broader than that of penicillin. Additionally, it exhibits antiviral activity against HIV-1.
  • Antibiotic useful for treating bacterial infections.
  • Antimicrobial spectrum similar to or slightly wider than penicillin.
  • Has antiviral activity against HIV-1.
  • For research use only.