Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythromycin Ethylsuccinate, G-Biosciences
SCI_ED

Catalog No. S139803
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139803 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139803 Supplier G-Biosciences Supplier No. RC1013
Add to Cart
Add to Cart

Erythromycin ethylsuccinate is a macrolide antibiotic with a target spectrum similar to that of penicillin. It has shown high efficacy against Mycoplasma pneumoniae and Legionella pneumophila.

it has been used to study erythromycin-resistance bacteria, down-regulation of motilin receptors, and mutations on the omsA and firA genes.

Chemical Identifiers

CAS 1264-62-6

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 25 g
Percent Purity 98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.