Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythromycin, G-Biosciences
SCI_ED

Catalog No. S139802
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139802 25 g
S139719 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139802 Supplier G-Biosciences Supplier No. RC1010
Add to Cart
Add to Cart

Erythromycin is a macrolide antibiotic isolated from Streptomyces erythreus. It displays strong efficacy against gram-positive cocci and bacilli and some mycobacteria, mycoplasma, and spirochetes.

In addition to its antimicrobial properties, erythromycin is a motilin receptor agonist that has been shown to inhibit respiratory glycoconjugate secretion in vitro. It has also been used to select for recombinant Lactococcus lactis MG1363 strains in microbiological applications.

Chemical Identifiers

CAS 114-07-8

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 25 g
Percent Purity 98%
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.