Learn More
Medchemexpress LLC Erythromycin lactobionate | 3847-29-8 | 100.0% | 25 MG
SDP
Catalog No. 5000450561
Encompass_Preferred
Erythromycin lactobionate is a macrolide antibiotic produced by the actinomycete *Streptomyces erythreus*. It exhibits a broad spectrum of antimicrobial activity by binding to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis. It has also demonstrated antitumor and neuroprotective effects in various research studies.
  • Macrolide antibiotic
  • Inhibits RNA-dependent protein synthesis
  • Exhibits antitumor and neuroprotective effects
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in H2O: ≥ 100 mg/mL (91.56 mM)
Catalog No. 50-004-50561 Supplier Medchemexpress LLC Supplier No. 25MGHYB0220A
Erythromycin lactobionate is a macrolide antibiotic produced by the actinomycete *Streptomyces erythreus*. It exhibits a broad spectrum of antimicrobial activity by binding to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis. It has also demonstrated antitumor and neuroprotective effects in various research studies.
  • Macrolide antibiotic
  • Inhibits RNA-dependent protein synthesis
  • Exhibits antitumor and neuroprotective effects
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in H2O: ≥ 100 mg/mL (91.56 mM)