Learn More
Medchemexpress LLC Erythromycin lactobionate | 3847-29-8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0220A50MG
Encompass_Preferred
Erythromycin lactobionate is a macrolide antibiotic produced by the actinomycete Streptomyces erythreus with a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits, inhibiting RNA-dependent protein synthesis without affecting nucleic acid synthesis. This compound also demonstrates antitumor and neuroprotective effects in research.
  • Macrolide antibiotic
  • Broad spectrum of antimicrobial activity
  • Inhibits bacterial protein synthesis
  • Exhibits antitumor effects
  • Exhibits neuroprotective effects
Catalog No. 50-004-05716