Learn More
Medchemexpress LLC Erythromycin lactobionate | 3847-29-8 | MFCD00868832 | 100 MG
SDP
Catalog No. 5000405434
Encompass_Preferred
Erythromycin lactobionate is a macrolide antibiotic produced by actinomycete *Streptomyces erythreus* with a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits and inhibits RNA-dependent protein synthesis by blockage of transpeptidation and/or translocation reactions, without affecting synthesis of nucleic acid. Erythromycin lactobionate also exhibits antitumor and neuroprotective effects in different fields of research.
  • Inhibits RNA-dependent protein synthesis
  • Exhibits antitumor effects
  • Provides neuroprotective benefits
  • Inhibits growth of *P. falciparum*
  • Shows antioxidant properties
  • Reduces inflammation by suppressing 4-HNE and 8-OHdG, and reducing Iba-1 and TNF-a expression
Catalog No. 50-004-05434 Supplier Medchemexpress LLC Supplier No. HYB0220A100MG
Erythromycin lactobionate is a macrolide antibiotic produced by actinomycete *Streptomyces erythreus* with a broad spectrum of antimicrobial activity. It binds to bacterial 50S ribosomal subunits and inhibits RNA-dependent protein synthesis by blockage of transpeptidation and/or translocation reactions, without affecting synthesis of nucleic acid. Erythromycin lactobionate also exhibits antitumor and neuroprotective effects in different fields of research.
  • Inhibits RNA-dependent protein synthesis
  • Exhibits antitumor effects
  • Provides neuroprotective benefits
  • Inhibits growth of *P. falciparum*
  • Shows antioxidant properties
  • Reduces inflammation by suppressing 4-HNE and 8-OHdG, and reducing Iba-1 and TNF-a expression