Learn More
Medchemexpress LLC Erythromycin (thiocyanate) | 7704-67-8 | 96.4% | 1 G
SDP
Catalog No. 5000407186
Encompass_Preferred
Erythromycin thiocyanate is a macrolide antibiotic produced by *Streptomyces erythreus*, known for its broad-spectrum antimicrobial activity. It functions by inhibiting bacterial 50S ribosomal subunits, thereby blocking RNA-dependent protein synthesis. This compound also exhibits antitumor and neuroprotective effects.
  • Inhibits RNA-dependent protein synthesis
  • Broad-spectrum antimicrobial activity
  • Antitumor effects
  • Neuroprotective effects
Catalog No. 50-004-07186 Supplier Medchemexpress LLC Supplier No. HYB0220D1G
Erythromycin thiocyanate is a macrolide antibiotic produced by *Streptomyces erythreus*, known for its broad-spectrum antimicrobial activity. It functions by inhibiting bacterial 50S ribosomal subunits, thereby blocking RNA-dependent protein synthesis. This compound also exhibits antitumor and neuroprotective effects.
  • Inhibits RNA-dependent protein synthesis
  • Broad-spectrum antimicrobial activity
  • Antitumor effects
  • Neuroprotective effects