Learn More
Medchemexpress LLC Erythromycin thiocyanate | 7704-67-8 | 96.4% | 1 ML
SDP
Catalog No. 5000405717
Encompass_Preferred
Erythromycin thiocyanate, a macrolide antibiotic from Streptomyces erythreus, offers broad-spectrum antimicrobial activity. It inhibits bacterial 50S ribosomal subunits, blocking protein synthesis without affecting nucleic acid synthesis. The compound also demonstrates antitumor and neuroprotective effects in various research applications.
  • Macrolide antibiotic
  • Broad-spectrum antimicrobial activity
  • Inhibits bacterial protein synthesis by targeting 50S ribosomal subunits
  • Demonstrates antitumor effects
  • Provides neuroprotective effects
  • For research use only
Catalog No. 50-004-05717 Supplier Medchemexpress LLC Supplier No. HYB0220D10MM/1ML
Erythromycin thiocyanate, a macrolide antibiotic from Streptomyces erythreus, offers broad-spectrum antimicrobial activity. It inhibits bacterial 50S ribosomal subunits, blocking protein synthesis without affecting nucleic acid synthesis. The compound also demonstrates antitumor and neuroprotective effects in various research applications.
  • Macrolide antibiotic
  • Broad-spectrum antimicrobial activity
  • Inhibits bacterial protein synthesis by targeting 50S ribosomal subunits
  • Demonstrates antitumor effects
  • Provides neuroprotective effects
  • For research use only