Learn More
Selleck Chemical LLC Erythromycin thiocyanate S5227-25mg
SDP
Catalog No. 503111995
Encompass_Preferred
Erythromycin is a macrolide antibiotic that inhibits bacterial protein synthesis by binding to bacterial 50S ribosomal subunits and inhibits peptidyl transferase activity and interferes with translocation of amino acids during translation and assembly of proteins
Catalog No. 50-311-1995 Supplier Selleck Chemical LLC Supplier No. S522725MG
Erythromycin is a macrolide antibiotic that inhibits bacterial protein synthesis by binding to bacterial 50S ribosomal subunits and inhibits peptidyl transferase activity and interferes with translocation of amino acids during translation and assembly of proteins