Learn More
Cayman Chemical ErythromycylamIn 10mg
SDP
Catalog No. 502939584
Encompass_Preferred
A macrolide antibiotic; active against a variety of bacteria, including strains of S. pyogenes, S. pneumoniae, L. monocytogenes, and B. pertussis (MICs = 0.06-0.12, 0.06-0.12, 1-2, and 0.015-0.25 UG/ml, respectively); active against 28 clinical isolates of MAC complex isolated from patients with AIDS (MICs = 4-16 UG/ml); inhibits polylysine and polyproline synthesis in a cell-free assay
Catalog No. 50-293-9584 Supplier Cayman Chemical Supplier No. 2809810 MG
A macrolide antibiotic; active against a variety of bacteria, including strains of S. pyogenes, S. pneumoniae, L. monocytogenes, and B. pertussis (MICs = 0.06-0.12, 0.06-0.12, 1-2, and 0.015-0.25 UG/ml, respectively); active against 28 clinical isolates of MAC complex isolated from patients with AIDS (MICs = 4-16 UG/ml); inhibits polylysine and polyproline synthesis in a cell-free assay