Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythromycylamine | 26116-56-3 | MFCD00872156 | 97.0% | 734.96 g/mol | C37H70N2O12 | 250 MG
SDP

Catalog No. 5000208472
Encompass_Preferred

Erythromycylamine is a macrolide antibiotic used for research that exhibits antibacterial activity against many Gram-positive cocci and Moraxella catarrhalis. It is supplied as a solid with a reported purity of 97.0% and demonstrates solubility in DMSO (≥ 15 mg/mL).

  • Macrolide antibiotic with broad activity against Gram-positive cocci and Moraxella catarrhalis.
  • Supplied as a solid suitable for in vitro research applications.
  • Reported purity: 97.0%.
  • Soluble in DMSO at ≥ 15 mg/mL; hygroscopic DMSO may affect solubility.
  • Molecular formula C37H70N2O12, molecular weight 734.96 g/mol.

Catalog No. 50-002-08472 Supplier Medchemexpress LLC Supplier No. 250MGHYW055872
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erythromycylamine is a macrolide antibiotic used for research that exhibits antibacterial activity against many Gram-positive cocci and Moraxella catarrhalis. It is supplied as a solid with a reported purity of 97.0% and demonstrates solubility in DMSO (≥ 15 mg/mL).

  • Macrolide antibiotic with broad activity against Gram-positive cocci and Moraxella catarrhalis.
  • Supplied as a solid suitable for in vitro research applications.
  • Reported purity: 97.0%.
  • Soluble in DMSO at ≥ 15 mg/mL; hygroscopic DMSO may affect solubility.
  • Molecular formula C37H70N2O12, molecular weight 734.96 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.