Learn More
Medchemexpress LLC Erythropoietin (recombinant human) | 11096-26-7 | 100UG
SDP
Catalog No. 5000174804
Encompass_Preferred
Recombinant human erythropoietin (EPO) produced in HEK293 mammalian cells. Supplied lyophilized and tag-free, this glycosylated cytokine is suitable for cell-based proliferation assays, receptor binding studies, and ELISA. Formulated in PBS with trehalose, it has low endotoxin and demonstrated biological activity in TF-1 assays.
Catalog No. 50-001-74804 Supplier Medchemexpress LLC Supplier No. HYP703756100UG
Recombinant human erythropoietin (EPO) produced in HEK293 mammalian cells. Supplied lyophilized and tag-free, this glycosylated cytokine is suitable for cell-based proliferation assays, receptor binding studies, and ELISA. Formulated in PBS with trehalose, it has low endotoxin and demonstrated biological activity in TF-1 assays.