Learn More
Medchemexpress LLC Erythropoietin/EPO 10ug
SDP
Catalog No. 5000195432
Encompass_Preferred
Recombinant human erythropoietin (EPO) expressed in CHO cells and supplied as a lyophilized powder formulated in PBS (pH 7.4). The protein is >95% pure by reducing SDS-PAGE, shows an apparent molecular weight of ~26-38 kDa due to glycosylation, and demonstrates biological activity in TF-1 cell proliferation assays with ED50 in the low ng/mL range. Supplied in small vial sizes for research use, including a 10 μg pack.
  • CHO-expressed recombinant human EPO suitable for in vitro research.
  • Lyophilized from PBS, pH 7.4, for convenient storage and handling.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight of ~26-38 kDa due to glycosylation.
  • Validated biological activity in TF-1 cell proliferation assays (ED50 in the low ng/mL range).
  • Stable at -20°C; aliquot and store at -80°C for extended storage.
Catalog No. 50-001-95432 Supplier Medchemexpress LLC Supplier No. HYP716410UG
Recombinant human erythropoietin (EPO) expressed in CHO cells and supplied as a lyophilized powder formulated in PBS (pH 7.4). The protein is >95% pure by reducing SDS-PAGE, shows an apparent molecular weight of ~26-38 kDa due to glycosylation, and demonstrates biological activity in TF-1 cell proliferation assays with ED50 in the low ng/mL range. Supplied in small vial sizes for research use, including a 10 μg pack.
  • CHO-expressed recombinant human EPO suitable for in vitro research.
  • Lyophilized from PBS, pH 7.4, for convenient storage and handling.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight of ~26-38 kDa due to glycosylation.
  • Validated biological activity in TF-1 cell proliferation assays (ED50 in the low ng/mL range).
  • Stable at -20°C; aliquot and store at -80°C for extended storage.