Learn More
Medchemexpress LLC Erythropoietin/EPO 20ug
SDP
Catalog No. 5000180905
Encompass_Preferred
This gene encodes the erythropoietin (EPO) protein that is critical for regulating erythropoiesis It is mainly expressed in the liver during fetal development and is transferred to the kidneys in adults Erythropoietin/EPO Protein Mouse (HEK293 solution) is the recombinant mouse-derived Erythropoietin/EPO protein expressed by HEK293 with tag free
Catalog No. 50-001-80905 Supplier Medchemexpress LLC Supplier No. HYP7303820UG
This gene encodes the erythropoietin (EPO) protein that is critical for regulating erythropoiesis It is mainly expressed in the liver during fetal development and is transferred to the kidneys in adults Erythropoietin/EPO Protein Mouse (HEK293 solution) is the recombinant mouse-derived Erythropoietin/EPO protein expressed by HEK293 with tag free