Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythropoietin (EPO) | 11096-26-7 | >95.0% | 20UG
SDP

Catalog No. 5000175525
Encompass_Preferred

Recombinant human erythropoietin (EPO) expressed in HEK293 cells, tag-free and supplied as a lyophilized powder. The protein retains mammalian glycosylation important for activity and is formulated from filtered PBS with trehalose stabilizer. It is suitable for biochemical assays, receptor binding, and cell-based erythropoiesis studies.

  • Expression system: HEK293 cells
  • Tag: tag-free
  • Amount: 20 μg (lyophilized)
  • Purity: >95.0% (by PAGE or SEC-HPLC)
  • Molecular weight: approximately 28-45 kDa (glycosylated)
  • Endotoxin: <1 EU/μg
  • Formulation: lyophilized from 0.22 μm filtered PBS with trehalose
  • Applications: biochemical assays and cell-based erythropoiesis studies

Catalog No. 50-001-75525 Supplier Medchemexpress LLC Supplier No. HYP70375620UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human erythropoietin (EPO) expressed in HEK293 cells, tag-free and supplied as a lyophilized powder. The protein retains mammalian glycosylation important for activity and is formulated from filtered PBS with trehalose stabilizer. It is suitable for biochemical assays, receptor binding, and cell-based erythropoiesis studies.

  • Expression system: HEK293 cells
  • Tag: tag-free
  • Amount: 20 μg (lyophilized)
  • Purity: >95.0% (by PAGE or SEC-HPLC)
  • Molecular weight: approximately 28-45 kDa (glycosylated)
  • Endotoxin: <1 EU/μg
  • Formulation: lyophilized from 0.22 μm filtered PBS with trehalose
  • Applications: biochemical assays and cell-based erythropoiesis studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.