Learn More
Medchemexpress LLC Erythropoietin (EPO) protein, human (HEK293) | 11096-26-7 | 50UG
SDP
Catalog No. 5000174721
Encompass_Preferred
Recombinant human erythropoietin produced in HEK293 cells, supplied as a lyophilized, tag-free protein for research use. The glycosylated protein corresponds to amino acids A28-R193 and demonstrates bioactivity in TF-1 cell proliferation assays (ED50 0.4-1 ng/mL).
Catalog No. 50-001-74721 Supplier Medchemexpress LLC Supplier No. HYP70375650UG
Add to Cart
Add to Cart
Recombinant human erythropoietin produced in HEK293 cells, supplied as a lyophilized, tag-free protein for research use. The glycosylated protein corresponds to amino acids A28-R193 and demonstrates bioactivity in TF-1 cell proliferation assays (ED50 0.4-1 ng/mL).