Learn More
Sigma Aldrich Fine Chemicals Biosciences Erythropoietin EPO human r
Catalog No. 502904276
Encompass
Recombinant human erythropoietin (EPO) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of ~18 kDa. This protein is manufactured in human cells using an all-human production system with full chemically defined ingredients and with no serum. The human cells expression system allows human-like glycosylation and folding and often supports better stability of the protein in culture. The protein is produced with no artificial tags.
Catalog No. 50-290-4276 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E554650UG
Recombinant human erythropoietin (EPO) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of ~18 kDa. This protein is manufactured in human cells using an all-human production system with full chemically defined ingredients and with no serum. The human cells expression system allows human-like glycosylation and folding and often supports better stability of the protein in culture. The protein is produced with no artificial tags.