Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erythropoietin protein, cynomolgus (HEK293, His) | 11096-26-7 | >95.0% | 50 UG
SDP

Catalog No. 5000293056
Encompass_Preferred

Recombinant cynomolgus erythropoietin produced in HEK293 cells and supplied lyophilized with a C-terminal His tag for research applications. Provided as a stable lyophilized powder formulated for storage and reconstitution suitable for receptor binding and functional assays.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal 10xHis tag for purification and detection.
  • Amino acid range A28-R192 representing the mature protein.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation from PBS with 5% trehalose, 5% mannitol, 0.01% Tween 80.
  • Molecular weight approximately 19.7 kDa.
  • Store at -20°C; aliquot and store at -80°C for extended preservation.

Catalog No. 50-002-93056 Supplier Medchemexpress LLC Supplier No. HYP7522250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cynomolgus erythropoietin produced in HEK293 cells and supplied lyophilized with a C-terminal His tag for research applications. Provided as a stable lyophilized powder formulated for storage and reconstitution suitable for receptor binding and functional assays.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal 10xHis tag for purification and detection.
  • Amino acid range A28-R192 representing the mature protein.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation from PBS with 5% trehalose, 5% mannitol, 0.01% Tween 80.
  • Molecular weight approximately 19.7 kDa.
  • Store at -20°C; aliquot and store at -80°C for extended preservation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.