Learn More
Medchemexpress LLC Erythropoietin recep 50ug
SDP
Catalog No. 5000180177
Encompass_Preferred
The EPOR Protein functions as a receptor for erythropoietin mediating erythroblast proliferation and differentiation upon EPO stimulation The heterodimer triggers the JAK2/STAT5 signaling cascade and in certain cell types activates STAT1 STAT3 and the LYN tyrosine kinase Additionally isoform EPOR-T acts as a dominant-negative receptor modulating EPOR-mediated signaling pathways Erythropoietin receptor/EpoR Protein Human (HEK293 Fc) is the recombinant human-derived Erythropoietin receptor/EpoR protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80177 Supplier Medchemexpress LLC Supplier No. HYP7352150UG
The EPOR Protein functions as a receptor for erythropoietin mediating erythroblast proliferation and differentiation upon EPO stimulation The heterodimer triggers the JAK2/STAT5 signaling cascade and in certain cell types activates STAT1 STAT3 and the LYN tyrosine kinase Additionally isoform EPOR-T acts as a dominant-negative receptor modulating EPOR-mediated signaling pathways Erythropoietin receptor/EpoR Protein Human (HEK293 Fc) is the recombinant human-derived Erythropoietin receptor/EpoR protein expressed by HEK293 with C-hFc labeled tag