Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrosin B

Catalog No. aaa1418014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418014 25 g
AAA1418022 100 g
AAA1418036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1418014 Supplier Thermo Scientific Chemicals Supplier No. A1418014
Only null left
Add to Cart

Erythrosin B, C40H14I8Na2O10, CAS Number-16423-68-0, C.I. 45430; Tetraiodofluorescein sodium salt, 500g, 240-046-0, 879.87, H302, GHS H Statement H302 Harmful if swallowed., P264b-P270-P301+P312-P330-P501c, DKRYETBOLGDIOK-UHFFFAOYSA-L | CAS: 16423-68-0 | C40H14I8Na2O10 | 1715.757 g/mol

Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

Solubility
Soluble in water, glycerin and methanol.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16423-68-0
MDL Number MFCD00144257
PubChem CID 131676154

Specifications

Molecular Formula C20H6I4Na2O5
Solubility Information Soluble in water,glycerin and methanol.
InChI Key IINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
IUPAC Name disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Weight (g/mol) 879.86
Formula Weight 879.87
Sensitivity Hygroscopic; Light sensitive
Quantity 25 g
Chemical Name or Material Erythrosin B
Hazard Category H302
Hazard Statement GHS H Statement
H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 240-046-0
RTECSNumber LM5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.