Learn More
Cayman Chemical ErythrosIn B 100g
SDP
Catalog No. 502991170
Encompass_Preferred
A red fluorescein dye and an inhibitor of dopamine reuptake (IC50 = 45 µM in rat brain synaptosomes); has been used in the colorimetric and fluorescent determination of protein concentrations in vitro
Catalog No. 50-299-1170 Supplier Cayman Chemical Supplier No. 31722100 G
A red fluorescein dye and an inhibitor of dopamine reuptake (IC50 = 45 µM in rat brain synaptosomes); has been used in the colorimetric and fluorescent determination of protein concentrations in vitro