Learn More
Medchemexpress LLC HY-D0259 100mg Medchemexpress, Erythrosin B CAS:16423-68-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0259100MG
Encompass_Preferred
Medchemexpress, HY-D0259 100mg Erythrosin B CAS:16423-68-0 Erythrosin B is an artificial dye widely used in the food and textile industries. Erythrosin B is also a novel photosensitizer which has been used to develop animal models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3166