Learn More
Cayman Chemical Erythrosin B, Purity: ≥85%, CAS: 16423-68-0, 250 g
SDP
Catalog No. 502992511
Encompass_Preferred
A red fluorescein dye and an inhibitor of dopamine reuptake (IC50 = 45 µM in rat brain synaptosomes); has been used in the colorimetric and fluorescent determination of protein concentrations in vitro
Catalog No. 50-299-2511 Supplier Cayman Chemical Supplier No. 31722250 G
A red fluorescein dye and an inhibitor of dopamine reuptake (IC50 = 45 µM in rat brain synaptosomes); has been used in the colorimetric and fluorescent determination of protein concentrations in vitro