Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Erythrosin B, Certified, C.I. DcEr-5 10 GR
SDP

Catalog No. 50980457
Encompass_Preferred

(Acid Red 51, Erythrosin extra Bluish; Eosin J; Iodeosin B; Pyrosin B)
CAS #16423-68-0 C₂₀H₆O₅I₄Na₂ F.W. 879.87
Solubility: 10.0% Water; 5.0% Alcohol; 7.0% Cellosolve; 8.25% Glycol; 0.0% Xylene

Catalog No. 50-980-457 Supplier Electron Microscopy Sciences Supplier No. 15010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Acid Red 51, Erythrosin extra Bluish; Eosin J; Iodeosin B; Pyrosin B)
CAS #16423-68-0 C₂₀H₆O₅I₄Na₂ F.W. 879.87
Solubility: 10.0% Water; 5.0% Alcohol; 7.0% Cellosolve; 8.25% Glycol; 0.0% Xylene

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.