Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Erythrosin B, pure, high purity, biological stain

Catalog No. AC189460250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC189460250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC189460250 Supplier Thermo Scientific Chemicals Supplier No. 189460250
  • $81.00
    $65.65 / Each of 1

    Save $15.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16423-68-0
Molecular Formula C20H6I4Na2O5
Molecular Weight (g/mol) 879.86
MDL Number MFCD00144257
InChI Key IINNWAYUJNWZRM-UHFFFAOYSA-L
Synonym Acid Red 51, C.I. 45430
PubChem CID 12961638
IUPAC Name disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12

Specifications

Melting Point >300.0°C
Dye Content 87% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Beilstein 19, II, 258
Merck Index 15, 3749
Color Bordeaux to Red
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Quantity 25 g
Physical Form Fine Powder
Formula Weight 879.85
Grade Pure
Chemical Name or Material Erythrosin B
Show More Show Less
What is the alternative to TrypanBlue for using it with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000?

The suitable alternative would be Erythrosin B (Cat. No. 409451000, A14180.36, 189460250 and J62619.22).

Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.