Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrosin B, Sigma-Aldrich

Catalog No. 01259046 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-046 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-046 Supplier MilliporeSigma Supelco Supplier No. 19826925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erythrosin B has been used as a dye in fluorescence lifetime imaging.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16423-68-0
Molecular Weight (g/mol) 879.86 g/mol

Specifications

Color Dark Red
Quantity 25 g
Percent Purity ≥85 %
Grade Certified Biological Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.