Learn More
Medchemexpress LLC Erythrosin B sodium salt | 568-63-8 | MFCD00144257 | 85.0% | 879.86 g/mol | C20H6I4Na2O5 | 5 MG
SDP
Catalog No. 5000283201
Encompass_Preferred
Erythrosin B sodium salt is an iodinated xanthene dye supplied as an orange-red, water-soluble solid used as a red colorant and staining reagent in biochemical assays, histology, and textile applications. The compound has formula C20H6I4Na2O5, a reported purity of 85%, and is provided in a small package for low-volume laboratory use.
  • Water-soluble iodinated xanthene dye suitable for staining and assays.
  • Provides a vivid orange-red color for visualization and tracing.
  • Available in a small 5 mg package for low-volume laboratory use.
  • Reported purity of 85% suitable for general laboratory applications.
  • Store sealed, away from moisture and light to maintain stability.
Catalog No. 50-002-83201 Supplier Medchemexpress LLC Supplier No. HYW2069115MG
Erythrosin B sodium salt is an iodinated xanthene dye supplied as an orange-red, water-soluble solid used as a red colorant and staining reagent in biochemical assays, histology, and textile applications. The compound has formula C20H6I4Na2O5, a reported purity of 85%, and is provided in a small package for low-volume laboratory use.
  • Water-soluble iodinated xanthene dye suitable for staining and assays.
  • Provides a vivid orange-red color for visualization and tracing.
  • Available in a small 5 mg package for low-volume laboratory use.
  • Reported purity of 85% suitable for general laboratory applications.
  • Store sealed, away from moisture and light to maintain stability.