Learn More
Cole-Parmer Atago 4210 ES-421 Palette-style digital refractometer, 0.0 to 10.0% salinity
Catalog No. NC1796541 Shop All Cole-Parmer Products
Encompass_Preferred
Fast and accurate concentration readings make these compact portable refractometers ideal for on-site use. A built-in thermistor provides automatic temperature compensation (ATC) so you dont have to struggle with temperature correction tables. Error messages indicate concentrations or temperatures that fall out of range, inadequate sample volume, standardization errors, or low-battery. Easily calibrate units using distilled water and the zero-adjustment button.
Catalog No. NC1796541 Supplier Cole-Parmer Supplier No. 0294061
Fast and accurate concentration readings make these compact portable refractometers ideal for on-site use. A built-in thermistor provides automatic temperature compensation (ATC) so you dont have to struggle with temperature correction tables. Error messages indicate concentrations or temperatures that fall out of range, inadequate sample volume, standardization errors, or low-battery. Easily calibrate units using distilled water and the zero-adjustment button.