Learn More
Expression Systems 293-ES cells, Frozen in ESF SFM, 20M cells/vial
SDP
Catalog No. NC0700076
Encompass_Preferred
Expression Systems offers 293 cell lines adapted to serum-free suspension culture in ESF SFM. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
293-ES cells are derived from the human embryonic kidney HEK 293 cell line. 293-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial or suspension culture.
Capable of producing high levels of AAV vectors
Capable of high levels of recombinant protein expression
Good growth in adherent or suspension culture Quality and performance testing
Organism: Homo sapiens, human
Tissue: Embryonic Kidney
Cell Type: Epithelial
Biosafety Level 2: Cells contain adenovirus. Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.
Catalog No. NC0700076 Supplier Expression Systems Supplier No. 94007F
Expression Systems offers 293 cell lines adapted to serum-free suspension culture in ESF SFM. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
293-ES cells are derived from the human embryonic kidney HEK 293 cell line. 293-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial or suspension culture.
Capable of producing high levels of AAV vectors
Capable of high levels of recombinant protein expression
Good growth in adherent or suspension culture Quality and performance testing
Organism: Homo sapiens, human
Tissue: Embryonic Kidney
Cell Type: Epithelial
Biosafety Level 2: Cells contain adenovirus. Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.