Learn More
Biocare Medical, LLC Biocare Medical Herpes Simplex Virus 1 & 2 Antibody, Polyclonal IgG, Research Use Only, 6.0 mL
SDP
Supplier:  Biocare Medical, LLC PP108AA
Encompass_Preferred
The BioCare Medical Herpes Simplex Virus 1 & 2 antibody is designed for specific and precise identification of HSV-1 and HSV-2 in research settings. This rabbit polyclonal IgG antibody effectively targets major viral envelope glycoproteins and core proteins without cross-reactivity to other viruses, ensuring reliable results in immunohistochemistry applications. Its 6.0 mL predilute format is optimized for use with formalin-fixed, paraffin-embedded tissues, providing a valuable tool for comprehensive viral localization studies.
Catalog No. NC1476126