Learn More
Medchemexpress LLC Esamisulpride (S)-amisulpride | 71675-92-8 | 99.0% | 369.48 g/mol | C17H27N3O4S | 100MG
SDP
Catalog No. 5000235835
Encompass_Preferred
Esamisulpride ((S)-amisulpride) is the S-enantiomer of amisulpride, a potent dopamine D2/D3 receptor antagonist with additional 5-HT7 antagonism. Supplied as a solid for research use, it is used in pharmacology and neuroscience studies to investigate dopaminergic and serotonergic signaling. Molecular formula: C17H27N3O4S; molecular weight: 369.48 g/mol; CAS: 71675-92-8; typical purity ~99%.
  • High purity suitable for research use.
  • Stereochemically defined S-enantiomer for selective pharmacological studies.
  • Potent antagonist at dopamine D2/D3 and 5-HT7 receptors.
  • Supplied as a solid with recommended storage for powder at -20°C and solutions at -80°C.
  • Available in multiple pack sizes including 100 mg for laboratory experiments.
Catalog No. 50-002-35835 Supplier Medchemexpress LLC Supplier No. HY126068100MG
Esamisulpride ((S)-amisulpride) is the S-enantiomer of amisulpride, a potent dopamine D2/D3 receptor antagonist with additional 5-HT7 antagonism. Supplied as a solid for research use, it is used in pharmacology and neuroscience studies to investigate dopaminergic and serotonergic signaling. Molecular formula: C17H27N3O4S; molecular weight: 369.48 g/mol; CAS: 71675-92-8; typical purity ~99%.
  • High purity suitable for research use.
  • Stereochemically defined S-enantiomer for selective pharmacological studies.
  • Potent antagonist at dopamine D2/D3 and 5-HT7 receptors.
  • Supplied as a solid with recommended storage for powder at -20°C and solutions at -80°C.
  • Available in multiple pack sizes including 100 mg for laboratory experiments.