Learn More
Medchemexpress LLC Esamisulpride (S)-amisulpride | 71675-92-8 | 99.0% | 369.48 g/mol | C17H27N3O4S | 10MG
SDP
Catalog No. 5000235541
Encompass_Preferred
Esamisulpride (S)-amisulpride is the S-enantiomer of amisulpride supplied for research use as a dopamine D2/D3 receptor antagonist with low-nanomolar affinity. It also shows activity at 5-HT7 and is used in pharmacology and behavioral studies related to schizophrenia and depression.
  • High chemical purity (99.04%) suitable for research applications.
  • Potent D2 and D3 receptor antagonist with low-nanomolar affinity.
  • Reported 5-HT7 receptor activity for broader pharmacological profiling.
  • Available in small mass quantities appropriate for preclinical experiments.
  • Confirmed molecular formula C17H27N3O4S and molecular weight 369.48 g/mol.
Catalog No. 50-002-35541 Supplier Medchemexpress LLC Supplier No. HY12606810MG
Esamisulpride (S)-amisulpride is the S-enantiomer of amisulpride supplied for research use as a dopamine D2/D3 receptor antagonist with low-nanomolar affinity. It also shows activity at 5-HT7 and is used in pharmacology and behavioral studies related to schizophrenia and depression.
  • High chemical purity (99.04%) suitable for research applications.
  • Potent D2 and D3 receptor antagonist with low-nanomolar affinity.
  • Reported 5-HT7 receptor activity for broader pharmacological profiling.
  • Available in small mass quantities appropriate for preclinical experiments.
  • Confirmed molecular formula C17H27N3O4S and molecular weight 369.48 g/mol.