Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Esamisulpride (S)-amisulpride | 71675-92-8 | 99.0% | 369.48 g/mol | C17H27N3O4S | 25MG
SDP

Catalog No. 5000238312
Encompass_Preferred

Esamisulpride (S-amisulpride) is the S-enantiomer of amisulpride supplied as a research-grade chemical standard for pharmacology and receptor-binding studies. It acts as a dopamine D2/D3 receptor antagonist and is provided as a solid for in vitro receptor and pharmacological assays.

  • High purity: 99.0% (as stated by supplier).
  • Molecular formula: C17H27N3O4S; molecular weight: 369.48 g/mol.
  • CAS number: 71675-92-8.
  • Available in a 25 mg quantity suitable for small-scale research use.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solution at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Appearance: solid, white to light yellow.

Catalog No. 50-002-38312 Supplier Medchemexpress LLC Supplier No. HY12606825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Esamisulpride (S-amisulpride) is the S-enantiomer of amisulpride supplied as a research-grade chemical standard for pharmacology and receptor-binding studies. It acts as a dopamine D2/D3 receptor antagonist and is provided as a solid for in vitro receptor and pharmacological assays.

  • High purity: 99.0% (as stated by supplier).
  • Molecular formula: C17H27N3O4S; molecular weight: 369.48 g/mol.
  • CAS number: 71675-92-8.
  • Available in a 25 mg quantity suitable for small-scale research use.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solution at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Appearance: solid, white to light yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.