Learn More
Medchemexpress LLC Esamisulpride (S)-amisulpride | 71675-92-8 | 99.0% | 369.48 g/mol | C17H27N3O4S | 50MG
SDP
Catalog No. 5000237998
Encompass_Preferred
Esamisulpride is the S-enantiomer of amisulpride and acts as a potent dopamine D2/D3 receptor antagonist with additional 5-HT7 activity. It is supplied as a research chemical for in vitro and in vivo studies related to schizophrenia and depression, offered in multiple pack sizes and accompanied by analytical documentation.
  • Potent dopamine D2/D3 receptor antagonist with 5-HT7 activity.
  • High purity suitable for research use.
  • Available in multiple pack sizes, including 50 mg.
  • Provided with analytical documents such as COA, SDS, HNMR, and LCMS.
  • Stable under recommended storage conditions for powder and solutions.
Catalog No. 50-002-37998 Supplier Medchemexpress LLC Supplier No. HY12606850MG
Esamisulpride is the S-enantiomer of amisulpride and acts as a potent dopamine D2/D3 receptor antagonist with additional 5-HT7 activity. It is supplied as a research chemical for in vitro and in vivo studies related to schizophrenia and depression, offered in multiple pack sizes and accompanied by analytical documentation.
  • Potent dopamine D2/D3 receptor antagonist with 5-HT7 activity.
  • High purity suitable for research use.
  • Available in multiple pack sizes, including 50 mg.
  • Provided with analytical documents such as COA, SDS, HNMR, and LCMS.
  • Stable under recommended storage conditions for powder and solutions.