Learn More
Cayman Chemical Esaxerenone, Purity: ≥98%, CAS: 1632006-28-0, 50 mg
SDP
Catalog No. 502942853
Encompass_Preferred
A non-steroidal mineralocorticoid receptor antagonist (IC50 = 9.4 nM); selective for mineralocorticoid receptors over glucocorticoid, androgen, and progesterone receptors (IC50s = >10,000 nM for all); increases the urinary ratio of sodium to potassium in bilateral adrenalectomized rats at 0.3, 1, or 3 mg/kg; decreases systolic blood pressure in DOCA-salt hypertensive rats at 3 mg/kg; reduces systolic blood pressure, urinary protein levels, blood levels of BNP, kidney and left ventricle weights, and glomerulosclerosis, as well as increases blood potassium levels, in Dahl salt-sensitive rats fed a high-salt diet at 2 mg/kg per day
Catalog No. 50-294-2853 Supplier Cayman Chemical Supplier No. 4108250 MG
A non-steroidal mineralocorticoid receptor antagonist (IC50 = 9.4 nM); selective for mineralocorticoid receptors over glucocorticoid, androgen, and progesterone receptors (IC50s = >10,000 nM for all); increases the urinary ratio of sodium to potassium in bilateral adrenalectomized rats at 0.3, 1, or 3 mg/kg; decreases systolic blood pressure in DOCA-salt hypertensive rats at 3 mg/kg; reduces systolic blood pressure, urinary protein levels, blood levels of BNP, kidney and left ventricle weights, and glomerulosclerosis, as well as increases blood potassium levels, in Dahl salt-sensitive rats fed a high-salt diet at 2 mg/kg per day