Learn More
Medchemexpress LLC HY-100471 1mg Medchemexpress, Esaxerenone CAS:1632006-28-0 Purity:>98%
SDP
Catalog No. 501931847
Encompass_Preferred
Medchemexpress, HY-100471 1mg Esaxerenone CAS:1632006-28-0 Esaxerenone is a novel, highly potent and selective non-steroidal mineralocorticoid receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1847 Supplier Medchemexpress LLC Supplier No. HY1004711MG
Medchemexpress, HY-100471 1mg Esaxerenone CAS:1632006-28-0 Esaxerenone is a novel, highly potent and selective non-steroidal mineralocorticoid receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.