Learn More
Selleck Chemical LLC ESBA 105-A3098-1MG-5
SDP
Catalog No. 503139138
Encompass_Preferred
ESBA 105 is an antibody targeting tumor necrosis factor (TNF) It has an efficient intraocular penetration to anterior and posterior region of eye MW 145 14 KD
Catalog No. 50-313-9138 Supplier Selleck Chemical LLC Supplier No. A30981MG5
ESBA 105 is an antibody targeting tumor necrosis factor (TNF) It has an efficient intraocular penetration to anterior and posterior region of eye MW 145 14 KD