Learn More
Fine Science Tools ESCHENBACH MAGNIFIER SPECTACLE
SDP
Catalog No. NC9968459
Encompass
Product: Eschenbach Magnifier Spectacles with Band/ Weight: 70g/ Magnification: 2.5x/ Working Distance: 35cm/ Features: Full adjustments of almost all parts/ Unit: Each
This item is not returnable. View return policy
Catalog No. NC9968459 Supplier Fine Science Tools Supplier No. 2804030
Product: Eschenbach Magnifier Spectacles with Band/ Weight: 70g/ Magnification: 2.5x/ Working Distance: 35cm/ Features: Full adjustments of almost all parts/ Unit: Each