Learn More
ATCC Escherichia coli 045; Strain: 04-3529
Supplier:  ATCC BAA2649
Encompass
Escherichia coli strain 595072 is a whole-genome sequenced bacterium that has applications in enteric disease research, food testing, and infectious disease research. This strain is non-toxigenic and is serotype O45:H10.
Catalog No. 50-238-4201