Learn More
ATCC Escherichia coli
Supplier: ATCC 11229
Escherichia coli strain AMC 198 is a whole-genome sequenced strain with applications in testing membrane filters, absorbant pads, antimicrobial handwashing formulations, disinfectants, ink grids, and sanitizers. This product is also available as a certified reference material (ATCC CRM-11229).