Learn More
ATCC Escherichia coli
Supplier:  ATCC BAA3170
Encompass
Escherichia coli strain AR-0349 was isolated from a human rectal swab. This strain is cited as mcr-1 positive and exhibits resistance to Colistin and Polymyxin B.
Catalog No. 50-238-4648