Learn More
ATCC Escherichia coli; Strain CDC; B1409-C1
Supplier:  ATCC 43889
Encompass
Escherichia coli strain CDC B1409-C1 is a whole-genome sequenced bacterium that was isolated from the feces of a patient with hemolytic uremic syndrome in North Carolina. This bacterium is serotype O157:H7 and produces Shiga-like toxin II. It can be used as a quality control strain for Biosynth products.
Catalog No. 50-237-9883