Learn More
ATCC Clostridium cellulolyticum
Catalog No. 502380721
Encompass
Type strain. Degrades glucose and cellulose.
Catalog No. 50-238-0721 Supplier ATCC Supplier No. 35319
Type strain. Degrades glucose and cellulose.